accessibility menu, dialog, popup

UserName

CAS RN 16940-81-1

HFPFFFFF

CAS RN 16940-81-1

IUPAC Name: hydrogen hexafluoro-λ⁵-phosphanuide
Synonyms: hexafluorophosphoric acid
Molecular Weight (g/mol): 145.97
Molecular Formula: F6HP
InChi Key: LJQLCJWAZJINEB-UHFFFAOYSA-O
SMILES: [H+].F[P-](F)(F)(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)