missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexafluoroglutaric Acid 97.0+%, TCI America™
$926.52 - $926.52
Chemical Identifiers
| CAS | 376-73-8 |
|---|---|
| Molecular Formula | C5H2F6O4 |
| Molecular Weight (g/mol) | 240.06 |
| MDL Number | MFCD00004172 |
| InChI Key | CCUWGJDGLACFQT-UHFFFAOYSA-N |
| Synonym | hexafluoroglutaric acid, perfluoroglutaric acid, hexafluoropentanedioic acid, pentanedioic acid, hexafluoro, glutaric acid, hexafluoro, pentanedioic acid, 2,2,3,3,4,4-hexafluoro, acmc-209iut, perfluoropentanedioic acid, glutaric acid, hexafluoro-8ci |
| PubChem CID | 67827 |
| IUPAC Name | hexafluoropentanedioic acid |
| SMILES | OC(=O)C(F)(F)C(F)(F)C(F)(F)C(O)=O |
Chemical Identifiers
| 376-73-8 | |
| 240.06 | |
| CCUWGJDGLACFQT-UHFFFAOYSA-N | |
| 67827 | |
| OC(=O)C(F)(F)C(F)(F)C(F)(F)C(O)=O |
| C5H2F6O4 | |
| MFCD00004172 | |
| hexafluoroglutaric acid, perfluoroglutaric acid, hexafluoropentanedioic acid, pentanedioic acid, hexafluoro, glutaric acid, hexafluoro, pentanedioic acid, 2,2,3,3,4,4-hexafluoro, acmc-209iut, perfluoropentanedioic acid, glutaric acid, hexafluoro-8ci | |
| hexafluoropentanedioic acid |
Specifications
| 376-73-8 | |
| Red-White | |
| MFCD00004172 | |
| hexafluoroglutaric acid, perfluoroglutaric acid, hexafluoropentanedioic acid, pentanedioic acid, hexafluoro, glutaric acid, hexafluoro, pentanedioic acid, 2,2,3,3,4,4-hexafluoro, acmc-209iut, perfluoropentanedioic acid, glutaric acid, hexafluoro-8ci | |
| OC(=O)C(F)(F)C(F)(F)C(F)(F)C(O)=O | |
| 240.06 | |
| 240.06 | |
| Crystalline Powder |
| 97°C | |
| C5H2F6O4 | |
| 25 g | |
| CCUWGJDGLACFQT-UHFFFAOYSA-N | |
| hexafluoropentanedioic acid | |
| 67827 | |
| ≥97.0% (T) | |
| Hexafluoroglutaric Acid |
Safety and Handling
TSCA : Yes