accessibility menu, dialog, popup

UserName

CAS RN 376-73-8

IUPAC Name: hexafluoropentanedioic acid
Synonyms: perfluoroglutaric acid
Molecular Weight (g/mol): 240.06
Molecular Formula: C5H2F6O4
InChi Key: CCUWGJDGLACFQT-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)