missing translation for 'onlineSavingsMsg'
Learn More

Hexafluoroglutaric acid, 98%

Catalog No. AAA1784814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1784814 25 g
AAA1784806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1784814 Supplier Thermo Scientific Chemicals Supplier No. A1784814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 376-73-8 | C5H2F6O4 | 240.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 376-73-8
Molecular Formula C5H2F6O4
Molecular Weight (g/mol) 240.06
MDL Number MFCD00004172
InChI Key CCUWGJDGLACFQT-UHFFFAOYSA-N
Synonym hexafluoroglutaric acid, perfluoroglutaric acid, hexafluoropentanedioic acid, pentanedioic acid, hexafluoro, glutaric acid, hexafluoro, pentanedioic acid, 2,2,3,3,4,4-hexafluoro, acmc-209iut, perfluoropentanedioic acid, glutaric acid, hexafluoro-8ci
PubChem CID 67827
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(O)=O

Specifications

Melting Point 92°C to 97°C
Boiling Point 134°C to 138°C (3 mmHg)
Quantity 25 g
UN Number UN3261
Beilstein 1801049
Sensitivity Hygroscopic
IUPAC Name 2,2,3,3,4,4-hexafluoropentanedioic acid
Formula Weight 240.06
Percent Purity 98%
Chemical Name or Material Hexafluoroglutaric acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 206-813-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only