missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexafluorobenzene 99.0+%, TCI America™
$55.92 - $816.46
Chemical Identifiers
| CAS | 392-56-3 |
|---|---|
| Molecular Formula | C6F6 |
| Molecular Weight (g/mol) | 186.056 |
| MDL Number | MFCD00000288 |
| InChI Key | ZQBFAOFFOQMSGJ-UHFFFAOYSA-N |
| Synonym | hexafluorobenzene, perfluorobenzene, benzene, hexafluoro, hexafluorbenzol, unii-cmc18t611k, benzene, 1,2,3,4,5,6-hexafluoro, hexa fluorobenzene, hexafluoro benzene, pubchem18879, acmc-1bmus |
| PubChem CID | 9805 |
| ChEBI | CHEBI:38589 |
| IUPAC Name | 1,2,3,4,5,6-hexafluorobenzene |
| SMILES | C1(=C(C(=C(C(=C1F)F)F)F)F)F |
Chemical Identifiers
| 392-56-3 | |
| 186.056 | |
| ZQBFAOFFOQMSGJ-UHFFFAOYSA-N | |
| 9805 | |
| 1,2,3,4,5,6-hexafluorobenzene |
| C6F6 | |
| MFCD00000288 | |
| hexafluorobenzene, perfluorobenzene, benzene, hexafluoro, hexafluorbenzol, unii-cmc18t611k, benzene, 1,2,3,4,5,6-hexafluoro, hexa fluorobenzene, hexafluoro benzene, pubchem18879, acmc-1bmus | |
| CHEBI:38589 | |
| C1(=C(C(=C(C(=C1F)F)F)F)F)F |
Specifications
| 392-56-3 | |
| Yellow | |
| C6F6 | |
| 5 g | |
| hexafluorobenzene, perfluorobenzene, benzene, hexafluoro, hexafluorbenzol, unii-cmc18t611k, benzene, 1,2,3,4,5,6-hexafluoro, hexa fluorobenzene, hexafluoro benzene, pubchem18879, acmc-1bmus | |
| C1(=C(C(=C(C(=C1F)F)F)F)F)F | |
| 186.056 | |
| CHEBI:38589 | |
| ≥99.0% (GC) | |
| Hexafluorobenzene |
| 5°C | |
| 81°C | |
| MFCD00000288 | |
| 1993 | |
| ZQBFAOFFOQMSGJ-UHFFFAOYSA-N | |
| 1,2,3,4,5,6-hexafluorobenzene | |
| 9805 | |
| 186.06 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0075
RTECSNumber : DA3050000
TSCA : Yes