missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexafluorobenzene 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 5°C |
| Color | Yellow |
| Boiling Point | 81°C |
| Quantity | 25 g |
| UN Number | 1993 |
| Formula Weight | 186.06 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Hexafluorobenzene |
Safety and Handling
| EINECSNumber | (3)-0075 |
| RTECSNumber | DA3050000 |
| TSCA | Yes |