missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexafluoroacetone Hydrate 97.0+%, TCI America™
$42.87 - $87.60
Chemical Identifiers
| CAS | 34202-69-2 |
|---|---|
| Molecular Formula | C3H6F6O4 |
| Molecular Weight (g/mol) | 220.067 |
| MDL Number | MFCD00149061 |
| InChI Key | SNZAEUWCEHDROX-UHFFFAOYSA-N |
| Synonym | hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate |
| PubChem CID | 36719 |
| IUPAC Name | 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate |
| SMILES | C(=O)(C(F)(F)F)C(F)(F)F.O.O.O |
Chemical Identifiers
| 34202-69-2 | |
| 220.067 | |
| SNZAEUWCEHDROX-UHFFFAOYSA-N | |
| 36719 | |
| C(=O)(C(F)(F)F)C(F)(F)F.O.O.O |
| C3H6F6O4 | |
| MFCD00149061 | |
| hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate | |
| 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate |
Specifications
| 34202-69-2 | |
| C3H6F6O4 | |
| 5 g | |
| hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate | |
| C(=O)(C(F)(F)F)C(F)(F)F.O.O.O | |
| 220.067 | |
| 166.02 | |
| Liquid |
| Yellow | |
| MFCD00149061 | |
| 2552 | |
| SNZAEUWCEHDROX-UHFFFAOYSA-N | |
| 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate | |
| 36719 | |
| ≥97.0% (T) | |
| Hexafluoroacetone Hydrate |
Safety and Handling
EINECSNumber : (2)-0581
RTECSNumber : UC2700000
TSCA : No