Learn More
Hexaethylphosphorus Triamide, 95%, Thermo Scientific™
Chemical Identifiers
| CAS | 2283-11-6 |
|---|---|
| Molecular Formula | C12H30N3P |
| Molecular Weight (g/mol) | 247.36 |
| MDL Number | MFCD00009041 |
| InChI Key | FDIOSTIIZGWENY-UHFFFAOYSA-N |
| Synonym | tris diethylamino phosphine, hexaethylphosphorous triamide, hexaethyl phosphorous triamide, phosphorous triamide, hexaethyl, n,n,n',n',n,n-hexaethylphosphinetriamine, tris n,n-diethylamino phosphine, hexaethyltriaminophosphine, hexaethylphosphoruo triamide, hexaethylphosphorus triamide, n-bis diethylamino phosphanyl-n-ethylethanamine |
| PubChem CID | 75292 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC423080250
|
N/A | 25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC423081000
|
N/A | 100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C12H30N3P | |
| MFCD00009041 | |
| tris diethylamino phosphine, hexaethylphosphorous triamide, hexaethyl phosphorous triamide, phosphorous triamide, hexaethyl, n,n,n',n',n,n-hexaethylphosphinetriamine, tris n,n-diethylamino phosphine, hexaethyltriaminophosphine, hexaethylphosphoruo triamide, hexaethylphosphorus triamide, n-bis diethylamino phosphanyl-n-ethylethanamine |
Specifications
| 2283-11-6 | |
| 111°C (6.0mmHg) | |
| Authentic | |
| C12H30N3P | |
| ((C2H5)2N)3P | |
| 0.89 | |
| Solubility in water: insoluble | |
| 247.36 | |
| 247.36 | |
| Hexaethylphosphorus triamide |
| 0.8900g/mL | |
| 58°C | |
| 94% min. (GC) | |
| 1.4720 to 1.475 | |
| MFCD00009041 | |
| tris diethylamino phosphine, hexaethylphosphorous triamide, hexaethyl phosphorous triamide, phosphorous triamide, hexaethyl, n,n,n',n',n,n-hexaethylphosphinetriamine, tris n,n-diethylamino phosphine, hexaethyltriaminophosphine, hexaethylphosphoruo triamide, hexaethylphosphorus triamide, n-bis diethylamino phosphanyl-n-ethylethanamine | |
| FDIOSTIIZGWENY-UHFFFAOYSA-N | |
| 75292 | |
| 95% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
GHS Signal Word: Warning
EINECSNumber : 218-920-8
RUO – Research Use Only