missing translation for 'onlineSavingsMsg'
Learn More

Hexaethylphosphorus Triamide, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 423080250

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC423080250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2283-11-6
247.36
FDIOSTIIZGWENY-UHFFFAOYSA-N
75292
C12H30N3P
MFCD00009041
tris diethylamino phosphine, hexaethylphosphorous triamide, hexaethyl phosphorous triamide, phosphorous triamide, hexaethyl, n,n,n',n',n,n-hexaethylphosphinetriamine, tris n,n-diethylamino phosphine, hexaethyltriaminophosphine, hexaethylphosphoruo triamide, hexaethylphosphorus triamide, n-bis diethylamino phosphanyl-n-ethylethanamine

Specifications

Hexaethylphosphorus triamide
0.8900g/mL
58°C
94% min. (GC)
C12H30N3P
((C2H5)2N)3P
MFCD00009041
tris diethylamino phosphine, hexaethylphosphorous triamide, hexaethyl phosphorous triamide, phosphorous triamide, hexaethyl, n,n,n',n',n,n-hexaethylphosphinetriamine, tris n,n-diethylamino phosphine, hexaethyltriaminophosphine, hexaethylphosphoruo triamide, hexaethylphosphorus triamide, n-bis diethylamino phosphanyl-n-ethylethanamine
FDIOSTIIZGWENY-UHFFFAOYSA-N
75292
95%
2283-11-6
111°C (6.0mmHg)
Authentic
Glass bottle
1.4720 to 1.475
25g
0.89
Solubility in water: insoluble
247.36
247.36
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly

GHS Signal Word: Warning

EINECSNumber : 218-920-8

SDS
Documents

Documents

Promotions

Promotions