missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexaethylene Glycol Monomethyl Ether 97.0+%, TCI America™
$180.97 - $1630.62
Chemical Identifiers
| CAS | 23601-40-3 |
|---|---|
| Molecular Formula | C13H28O7 |
| Molecular Weight (g/mol) | 296.36 |
| MDL Number | MFCD00045996 |
| InChI Key | FHHGCKHKTAJLOM-UHFFFAOYSA-N |
| Synonym | mPEG6-Alcohol |
| PubChem CID | 90207 |
| ChEBI | CHEBI:44752 |
| IUPAC Name | 2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol |
| SMILES | COCCOCCOCCOCCOCCOCCO |
Chemical Identifiers
| 23601-40-3 | |
| 296.36 | |
| FHHGCKHKTAJLOM-UHFFFAOYSA-N | |
| 90207 | |
| 2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol |
| C13H28O7 | |
| MFCD00045996 | |
| mPEG6-Alcohol | |
| CHEBI:44752 | |
| COCCOCCOCCOCCOCCOCCO |
Specifications
| 23601-40-3 | |
| 167°C | |
| MFCD00045996 | |
| mPEG6-Alcohol | |
| COCCOCCOCCOCCOCCOCCO | |
| 296.36 | |
| CHEBI:44752 | |
| ≥97.0% (GC) | |
| Hexaethylene Glycol Monomethyl Ether |
| Yellow | |
| C13H28O7 | |
| 1 g | |
| FHHGCKHKTAJLOM-UHFFFAOYSA-N | |
| 2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol | |
| 90207 | |
| 296.36 | |
| Liquid |
Safety and Handling
EINECSNumber : (7)-0097&(2)-0444
TSCA : Yes