accessibility menu, dialog, popup

UserName

H3COOOOOOOH

CAS RN 23601-40-3

IUPAC Name: 2,5,8,11,14,17-hexaoxanonadecan-19-ol
Synonyms: hexaethylene glycol monomethyl ether
Molecular Weight (g/mol): 296.36
Molecular Formula: C13H28O7
InChi Key: FHHGCKHKTAJLOM-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCOCCOCCO
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results