missing translation for 'onlineSavingsMsg'
Learn More

Hexaethylene Glycol Monomethyl Ether 97.0+%, TCI America™

Catalog No. H08081G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08081G 1 g
H08085G 5 g
H080825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H08081G Supplier TCI America Supplier No. H08081G
Only null left

Chemical Identifiers

CAS 23601-40-3
Molecular Formula C13H28O7
Molecular Weight (g/mol) 296.36
MDL Number MFCD00045996
InChI Key FHHGCKHKTAJLOM-UHFFFAOYSA-N
Synonym mPEG6-Alcohol
PubChem CID 90207
ChEBI CHEBI:44752
IUPAC Name 2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol
SMILES COCCOCCOCCOCCOCCOCCO

Specifications

Color Yellow
Boiling Point 167°C
Quantity 1 g
Formula Weight 296.36
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexaethylene Glycol Monomethyl Ether
EINECSNumber (7)-0097&(2)-0444
TSCA Yes