missing translation for 'onlineSavingsMsg'
Learn More

Hexaethyl phosphorous triamide, 97%

CAS: 2283-11-6 | C12H30N3P | 247.37 g/mol

$120.75 - $1117.44

Chemical Identifiers

CAS 2283-11-6
MDL Number MFCD00009041
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1249006
View Documents
Thermo Scientific Chemicals
A1249006
5 g
Each for $120.75
Only null left
 
AAA1249014
View Documents
Thermo Scientific Chemicals
A1249014
25 g
Each for $353.05
Only null left
 
AAA1249022
View Documents
Thermo Scientific Chemicals
A1249022
100 g
Each for $1,117.44
Only null left
 
Description

Description

Hexaethyl phosphorous triamide is used in the Barton syntheses of hindered alkenes, involving desulfurization of small-ring heterocycles with elimination of nitrogen or carbon dioxide. It is also used as a reagent for selective vicinal dehalogenation of halogenated alkanes to give perhalogenated alkenes. The method has potential for the synthesis of highly fluorinated and mixed fluoro-halo alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaethyl phosphorous triamide is used in the Barton syntheses of hindered alkenes, involving desulfurization of small-ring heterocycles with elimination of nitrogen or carbon dioxide. It is also used as a reagent for selective vicinal dehalogenation of halogenated alkanes to give perhalogenated alkenes. The method has potential for the synthesis of highly fluorinated and mixed fluoro-halo alkenes.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
Specifications

Chemical Identifiers

2283-11-6
MFCD00009041

Specifications

2283-11-6
245°C to 246°C
C8H4F3NO
MFCD00009041
UN1993
Air Sensitive
Insoluble in water.
C1=CC=C(C(=C1)C(F)(F)F)N=C=O
187.12
247.37
Hexaethyl phosphorous triamide
0.903
58°C (136°F)
1.4737
5 g
636187
2-trifluoromethyl phenyl isocyanate, 1-isocyanato-2-trifluoromethyl benzene, benzene, 1-isocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isocyanate, o-trifluoromethylphenylisocyanate, isocyanic acid, alpha,alpha,alpha-trifluoro-o-tolyl ester, o-trifluoromethyl phenyl isocyanate, 2-trifluoromethyl phenylisocyanate, 2-trifluoromethyl benzenisocyanate, isocyanic acid, .alpha.,.alpha.,.alpha.-trifluoro-o-tolyl ester
GZWGTVZRRFPVAS-UHFFFAOYSA-N
1-isocyanato-2-(trifluoromethyl)benzene
16794
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 218-920-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only