missing translation for 'onlineSavingsMsg'
Learn More

Hexaethyl phosphorous triamide, 97%

CAS: 2283-11-6 | C12H30N3P | 247.37 g/mol

Supplier:  Thermo Scientific Chemicals A1249014

 View more versions of this product

Catalog No. AAA1249014

Only null left
Explore available promotions

Description

Description

Hexaethyl phosphorous triamide is used in the Barton syntheses of hindered alkenes, involving desulfurization of small-ring heterocycles with elimination of nitrogen or carbon dioxide. It is also used as a reagent for selective vicinal dehalogenation of halogenated alkanes to give perhalogenated alkenes. The method has potential for the synthesis of highly fluorinated and mixed fluoro-halo alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaethyl phosphorous triamide is used in the Barton syntheses of hindered alkenes, involving desulfurization of small-ring heterocycles with elimination of nitrogen or carbon dioxide. It is also used as a reagent for selective vicinal dehalogenation of halogenated alkanes to give perhalogenated alkenes. The method has potential for the synthesis of highly fluorinated and mixed fluoro-halo alkenes.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
Specifications

Specifications

Hexaethyl phosphorous triamide
0.903
58°C (136°F)
1.4737
25 g
636187
tris diethylamino phosphine, hexaethylphosphorous triamide, hexaethyl phosphorous triamide, phosphorous triamide, hexaethyl, hexaethyltriaminophosphine, n,n,n',n',n,n-hexaethylphosphinetriamine, tris n,n-diethylamino phosphine, hexaethylphosphoruo triamide, hexaethylphosphorus triamide, phosphorous triamide, n,n,n',n',n,n-hexaethyl
FDIOSTIIZGWENY-UHFFFAOYSA-N
N-[bis(diethylamino)phosphanyl]-N-ethylethanamine
75292
97%
2283-11-6
245°C to 246°C
C12H30N3P
MFCD00009041
UN1993
Air Sensitive
Insoluble in water.
CCN(CC)P(N(CC)CC)N(CC)CC
247.37
247.37

Chemical Identifiers

2283-11-6
MFCD00009041
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 218-920-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only