missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyltrimethylammonium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. H07345G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07345G 5 g
H07341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07345G Supplier TCI America Supplier No. H07345G
Only null left

Chemical Identifiers

CAS 73257-08-6
Molecular Formula C19H42BF4N
Molecular Weight (g/mol) 371.356
MDL Number MFCD00144896
InChI Key ZYVAQFUOBZOOLY-UHFFFAOYSA-N
Synonym Cetyltrimethylammonium Tetrafluoroborate
PubChem CID 16213810
IUPAC Name hexadecyl(trimethyl)azanium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Color White
Quantity 5 g
UN Number 1759
Formula Weight 371.36
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecyltrimethylammonium Tetrafluoroborate
EINECSNumber (2)-0184&(9)-0795&(9)-1971
TSCA No