CAS RN 73257-08-6
CAS RN 73257-08-6
IUPAC Name:
hexadecyltrimethylazanium; tetrafluoroboranuide
Synonyms:
cetyl-trimethyl-ammonium tetrafluoroborate
Molecular Weight (g/mol):
371.36
Molecular Formula:
C19H42BF4N
InChi Key:
ZYVAQFUOBZOOLY-UHFFFAOYSA-N
SMILES:
F[B-](F)(F)F.CCCCCCCCCCCCCCCC[N+](C)(C)C
1
–
1
of
1
results
1
Hexadecyltrimethylammonium Tetrafluoroborate 98.0+%, TCI America™
CAS: 73257-08-6 Molecular Formula: C19H42BF4N Molecular Weight (g/mol): 371.356 MDL Number: MFCD00144896 InChI Key: ZYVAQFUOBZOOLY-UHFFFAOYSA-N Synonym: Cetyltrimethylammonium Tetrafluoroborate PubChem CID: 16213810 IUPAC Name: hexadecyl(trimethyl)azanium;tetrafluoroborate SMILES: [B-](F)(F)(F)F.CCCCCCCCCCCCCCCC[N+](C)(C)C