missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyltrimethylammonium hydroxide, 25% in methanol

Catalog No. AC411420500
Change view
Click to view available options
Quantity:
50 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411420500 50 g Glass bottle
AC411421000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC411420500 Supplier Thermo Scientific Chemicals Supplier No. 411420500
Only null left

CAS: 505-86-2 | C19H43NO | 301.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 505-86-2
Molecular Formula C19H43NO
Molecular Weight (g/mol) 301.56
MDL Number MFCD00041921
InChI Key WJLUBOLDZCQZEV-UHFFFAOYSA-M
Synonym hexadecyltrimethylammonium hydroxide, cetyltrimethylammonium hydroxide, unii-8q7d6895lv, n,n,n-trimethylhexadecan-1-aminium hydroxide, 1-hexadecanaminium, n,n,n-trimethyl-, hydroxide, cetrimide inn:ban:jan, n-hexadecyltrimethylammonium hydroxide, n,n,n-trimethyl-1-hexadecanaminiuhydroxide, 1-hexadecanaminium, n,n,n-trimethyl-, hydroxide 1:1, cetyl-trimethylammonium hydroxide
PubChem CID 68166
IUPAC Name hexadecyltrimethylazanium hydroxide
SMILES [OH-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Color Colorless to Yellow
Linear Formula CH3(CH2)15N(OH)(CH3)3
Concentration 23 to 27 w/w% (in Methanol)
Density 0.83g/mL
Formula Weight 301.55
Packaging Glass bottle
Quantity 50 g
Physical Form Solution
Chemical Name or Material Hexadecyltrimethylammonium hydroxide, 25%in Methanol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes damage to organs.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-022-4
TSCA TSCA

RUO – Research Use Only