missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecyltrimethylammonium Chloride, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
Specifications
Specifications
| Melting Point | 234°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (Argentometry) |
| Packaging | Plastic bottle |
| Linear Formula | CH3(CH2)15N(Cl)(CH3)3 |
| Quantity | 25 g |
| Solubility Information | Solubility in water: soluble |
| Formula Weight | 319.75 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Toxic in contact with skin. Causes severe skin burns and eye damage. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF SWALLOWED |
| EINECSNumber | 203-928-6 |
RUO – Research Use Only