missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyltrimethylammonium Bromide 98.0+%, TCI America™

Catalog No. p-7130323
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H008125G 25 g
H0081100G 100 g
H0081500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H008125G Supplier TCI America Supplier No. H008125G
Only null left

Chemical Identifiers

CAS 57-09-0
Molecular Formula C19H42BrN
Molecular Weight (g/mol) 364.46
MDL Number MFCD00011772
InChI Key LZZYPRNAOMGNLH-UHFFFAOYSA-M
Synonym cetrimonium bromide, cetyltrimethylammonium bromide, hexadecyltrimethylammonium bromide, ctab, centimide, cetab, bromat, ctmab, cee dee, cetavlon
PubChem CID 5974
ChEBI CHEBI:3567
IUPAC Name hexadecyltrimethylazanium bromide
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Melting Point 243°C
Color White
pH 5.0 to 7.0 (50g/L H2O soln.)
Quantity 25 g
UN Number 3077
Formula Weight 364.46
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecyltrimethylammonium Bromide
EINECSNumber (2)-0184&(9)-0795
RTECSNumber BQ7875000
TSCA Yes