missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecyltrimethylammonium Bromide, 99+%
CAS: 57-09-0 | C19H42BrN | 364.46 g/mol
Supplier: Thermo Scientific Chemicals 227160025
| Quantity | 2.5 kg |
|---|---|
| Packaging | Plastic Drum |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
| 57-09-0 | |
| 364.46 | |
| LZZYPRNAOMGNLH-UHFFFAOYSA-M | |
| CHEBI:3567 |
| C19H42BrN | |
| MFCD00011772 | |
| 5974 | |
| [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C |
Specifications
| Hexadecyltrimethylammonium bromide | |
| 57-09-0 | |
| 230.0°C | |
| 2.5 kg | |
| Authentic | |
| 99+% | |
| CH3(CH2)15N(CH3)3Br | |
| 04, IV, 819 | |
| 15, 2024 | |
| Solubility in water: 13g/L (20°C). Other solubilities: soluble in methanol (>200g/L at 20 °C), ethanol, insoluble in benzene, diethyl ether, chloroform, sparingly soluble in acetone | |
| [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C | |
| 364.46 | |
| CHEBI:3567 | |
| ≥99% |
| White | |
| Crystalline Powder | |
| >230°C | |
| 244°C | |
| Plastic Drum | |
| C19H42BrN | |
| MFCD00011772 | |
| 15,77 | |
| Cetrimonium bromide, Cetyltrimethylammonium bromide, CTABr | |
| LZZYPRNAOMGNLH-UHFFFAOYSA-M | |
| hexadecyl(trimethyl)azanium;bromide | |
| 5974 | |
| 364.45 |
Safety and Handling
Warning