missing translation for 'onlineSavingsMsg'
Learn More

Hexadecylsuccinic Anhydride 95.0+%, TCI America™

Catalog No. H007925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H007925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H007925G Supplier TCI America Supplier No. H007925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4200-91-3
Molecular Formula C20H36O3
Molecular Weight (g/mol) 324.505
MDL Number MFCD00014550
InChI Key GBBHWGRJHHNAGT-UHFFFAOYSA-N
PubChem CID 77868
IUPAC Name 3-hexadecyloxolane-2,5-dione
SMILES CCCCCCCCCCCCCCCCC1CC(=O)OC1=O

Specifications

Melting Point 75°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 324.51
Percent Purity ≥95.0% (GC,T)
Chemical Name or Material Hexadecylsuccinic Anhydride
EINECSNumber (2)-0851
TSCA Yes