accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 4200-91-3

IUPAC Name: 3-hexadecyloxolane-2,5-dione
Synonyms: 3-hexadecyloxolane-2,5-dione
Molecular Weight (g/mol): 324.51
Molecular Formula: C20H36O3
InChi Key: GBBHWGRJHHNAGT-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCCC1CC(=O)OC1=O
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results