missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecylpyridinium Bromide Hydrate 96.0+%, TCI America™
$50.33 - $378.40
Chemical Identifiers
| CAS | 140-72-7 |
|---|---|
| Molecular Formula | C21H38BrN |
| Molecular Weight (g/mol) | 384.446 |
| MDL Number | MFCD00011988 |
| InChI Key | DVBJBNKEBPCGSY-UHFFFAOYSA-M |
| Synonym | cetylpyridinium bromide, 1-hexadecylpyridinium bromide, hexadecylpyridinium bromide, cetapharm, hexadecylpyridine bromide, 1-hexadecylpyridin-1-ium bromide, bromocet, cetazolin, nitrogenol, seprisan |
| PubChem CID | 8816 |
| ChEBI | CHEBI:77052 |
| IUPAC Name | 1-hexadecylpyridin-1-ium;bromide |
| SMILES | CCCCCCCCCCCCCCCC[N+]1=CC=CC=C1.[Br-] |
Chemical Identifiers
| 140-72-7 | |
| 384.446 | |
| DVBJBNKEBPCGSY-UHFFFAOYSA-M | |
| 8816 | |
| 1-hexadecylpyridin-1-ium;bromide |
| C21H38BrN | |
| MFCD00011988 | |
| cetylpyridinium bromide, 1-hexadecylpyridinium bromide, hexadecylpyridinium bromide, cetapharm, hexadecylpyridine bromide, 1-hexadecylpyridin-1-ium bromide, bromocet, cetazolin, nitrogenol, seprisan | |
| CHEBI:77052 | |
| CCCCCCCCCCCCCCCC[N+]1=CC=CC=C1.[Br-] |
Specifications
| 140-72-7 | |
| 6 to 8 (5g/L H2O soln. 20°C) | |
| MFCD00011988 | |
| cetylpyridinium bromide, 1-hexadecylpyridinium bromide, hexadecylpyridinium bromide, cetapharm, hexadecylpyridine bromide, 1-hexadecylpyridin-1-ium bromide, bromocet, cetazolin, nitrogenol, seprisan | |
| CCCCCCCCCCCCCCCC[N+]1=CC=CC=C1.[Br-] | |
| 384.446 | |
| CHEBI:77052 | |
| ≥96.0% (HPLC,T) | |
| Hexadecylpyridinium Bromide Hydrate |
| White | |
| C21H38BrN | |
| 25 g | |
| DVBJBNKEBPCGSY-UHFFFAOYSA-M | |
| 1-hexadecylpyridin-1-ium;bromide | |
| 8816 | |
| 384.45 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3686&(9)-1979
RTECSNumber : UU4848000
TSCA : Yes