accessibility menu, dialog, popup

UserName

BrH3CN

CAS RN 140-72-7

IUPAC Name: 1-hexadecylpyridin-1-ium bromide
Synonyms: cetylpyridinium bromide
Molecular Weight (g/mol): 384.45
Molecular Formula: C21H38BrN
InChi Key: DVBJBNKEBPCGSY-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCCCCCC[N+]1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results