missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecylpyridinium Bromide Hydrate 96.0+%, TCI America™
Specifications
Specifications
| Color | White |
| pH | 6 to 8 (5g/L H2O soln. 20°C) |
| Quantity | 25 g |
| Formula Weight | 384.45 |
| Percent Purity | ≥96.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Hexadecylpyridinium Bromide Hydrate |
Safety and Handling
| EINECSNumber | (5)-3686&(9)-1979 |
| RTECSNumber | UU4848000 |
| TSCA | Yes |