missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt 98.0+%, TCI America™

Catalog No. p-7130960
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12835G 5 g
H128325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12835G Supplier TCI America Supplier No. H12835G
Only null left

Chemical Identifiers

CAS 2281-11-0
Molecular Formula C21H45NO3S
Molecular Weight (g/mol) 391.66
MDL Number MFCD00036911
InChI Key TUBRCQBRKJXJEA-UHFFFAOYSA-N
Synonym 3-n,n-dimethylpalmitylammonio propanesulfonate, zwittergent 3-16, 3-hexadecyldimethylammonio propane-1-sulfonate, palmityl sulfobetaine, n-hexadecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-hexadecyl dimethyl ammonio propane-1-sulfonate, 3-palmityldimethylammonio propanesulfonate, 3-hexadecyldimethylammonio propanesulfonate, 3-n,n-dimethylhexadecylammonio propanesulfonate, n-hexadecy-n,n-dimethyl-3-ammonio-1-propanesulfonate
PubChem CID 92176
IUPAC Name 3-(hexadecyldimethylazaniumyl)propane-1-sulfonate
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

Specifications

Melting Point 242°C
Color White
Quantity 5 g
Formula Weight 391.66
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt [for Biochemical Research]