accessibility menu, dialog, popup

UserName

H3CNH3CCH3SOOO

CAS RN 2281-11-0

IUPAC Name: 3-(hexadecyldimethylazaniumyl)propane-1-sulfonate
Synonyms: 3-(hexadecyldimethylammonio)propane-1-sulfonate
Molecular Weight (g/mol): 391.66
Molecular Formula: C21H45NO3S
InChi Key: TUBRCQBRKJXJEA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)