missing translation for 'onlineSavingsMsg'
Learn More

Hexadecane, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187418
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014560 5 mL
111014559 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014560 Supplier Merck Emd Millipore Supplier No. 522095ML
Only null left

Hexadecane can find applications as a sole carbon source for the growth of canals, which are supramolecular structures observed in the cell wall of Candida maltose.

Featured Industries: Petroleum

Specifications

CAS 544-76-3
Ignition Point 395°F
Vapor Pressure 1 mmHg (105.3°C)
Vapor Density 7.8 (vs. air)
Density 0.773 g/mL (at 25°C)
Flash Point 135°C
Physical Form Neat
Linear Formula CH3(CH2)14 CH3
MDL Number MFCD00008998
Melting Point 18°C
Boiling Point 287°C
Synonym n-Hexadecane; Cetane
Type Analytical Standard
Quantity 5 mL
Formula Weight 226.44
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less