missing translation for 'onlineSavingsMsg'
Learn More

Hexadecanamide 95.0+%, TCI America™

Catalog No. H006710G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H006710G 10 g
H006725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H006710G Supplier TCI America Supplier No. H006710G
Only null left

Chemical Identifiers

CAS 629-54-9
Molecular Formula C16H33NO
Molecular Weight (g/mol) 255.446
MDL Number MFCD00025534
InChI Key HSEMFIZWXHQJAE-UHFFFAOYSA-N
Synonym Palmitamide
PubChem CID 69421
ChEBI CHEBI:74475
IUPAC Name hexadecanamide
SMILES CCCCCCCCCCCCCCCC(=O)N

Specifications

Melting Point 106°C
Color White
Boiling Point 236°C
Quantity 10 g
Formula Weight 255.45
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecanamide
EINECSNumber (2)-0824
TSCA Yes