accessibility menu, dialog, popup

UserName

H3CNH2O

CAS RN 629-54-9

IUPAC Name: hexadecanamide
Synonyms: cetamide
Molecular Weight (g/mol): 255.45
Molecular Formula: C16H33NO
InChi Key: HSEMFIZWXHQJAE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results