missing translation for 'onlineSavingsMsg'
Learn More

Hexadecahydropyrene (mixture of isomers) 97.0+%, TCI America™

Catalog No. H15761G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H15761G Supplier TCI America Supplier No. H15761G
Only null left

Chemical Identifiers

CAS 2435-85-0
Molecular Formula C16H26
Molecular Weight (g/mol) 218.384
MDL Number MFCD00021664
InChI Key BYBPEZLZCGOWIS-UHFFFAOYSA-N
PubChem CID 75524
IUPAC Name 1,2,3,3a,4,5,5a,6,7,8,8a,9,10,10a,10b,10c-hexadecahydropyrene
SMILES C1CC2CCC3CCCC4C3C2C(C1)CC4

Specifications

Color Yellow
Quantity 1 g
Formula Weight 218.38
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexadecahydropyrene (mixture of isomers)