accessibility menu, dialog, popup

UserName

CAS RN 2435-85-0

IUPAC Name: hexadecahydropyrene
Synonyms: perhydropyrene
Molecular Weight (g/mol): 218.38
Molecular Formula: C16H26
InChi Key: BYBPEZLZCGOWIS-UHFFFAOYNA-N
SMILES: C1CC2CCC3CCCC4CCC(C1)C2C34

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results