missing translation for 'onlineSavingsMsg'
Learn More

Hexadecafluoroheptane (mixture of isomers) 80.0+%, TCI America™

Catalog No. P085110G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P085110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P085110G Supplier TCI America Supplier No. P085110G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 335-57-9
Molecular Formula C7F16
Molecular Weight (g/mol) 388.051
MDL Number MFCD00040339
InChI Key LGUZHRODIJCVOC-UHFFFAOYSA-N
Synonym hexadecafluoroheptane, perfluoroheptane, perfluoro-n-heptane, heptane, hexadecafluoro, perfluoroheptanes, unii-i23zvd1p1l, i23zvd1p1l, heptane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-hexadecafluoro, perfluoroheptane s, acmc-20akry
PubChem CID 9553
ChEBI CHEBI:38847
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-hexadecafluoroheptane
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 82°C
Quantity 10 g
Formula Weight 388.05
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexadecafluoroheptane (mixture of isomers)
EINECSNumber (2)-2366
RTECSNumber MJ0875000
TSCA Yes