missing translation for 'onlineSavingsMsg'
Learn More

Hexacosane, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187422
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014966 250 mg
111014965 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014966 Supplier Merck Emd Millipore Supplier No. 52183250MG
Only null left
Featured Industries: Petroleum

Specifications

CAS 630-01-3
Physical Form Neat
Linear Formula CH3(CH2)24 CH3
MDL Number MFCD00009354
Melting Point 55°C to 58°C; 55°C to 58°C
Type Analytical Standard
Quantity 250 mg
Formula Weight 366.71
Percent Purity ≥98% (GC)
Grade Analytical Standard