missing translation for 'onlineSavingsMsg'
Learn More

Hexachlorophene 98.0+%, TCI America™

Catalog No. M02195G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02195G 5 g
M02191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M02195G Supplier TCI America Supplier No. M02195G
Only null left

Chemical Identifiers

CAS 70-30-4
Molecular Formula C13H6Cl6O2
Molecular Weight (g/mol) 406.889
MDL Number MFCD00002171
InChI Key ACGUYXCXAPNIKK-UHFFFAOYSA-N
Synonym hexachlorophene, hexachlorofen, hexachlorophen, almederm, exofene, nabac, gamophene, hexophene, steraskin, tersaseptic
PubChem CID 3598
ChEBI CHEBI:5693
IUPAC Name 3,4,6-trichloro-2-[(2,3,5-trichloro-6-hydroxyphenyl)methyl]phenol
SMILES C1=C(C(=C(C(=C1Cl)Cl)CC2=C(C(=CC(=C2Cl)Cl)Cl)O)O)Cl

Specifications

Melting Point 163°C
Quantity 5 g
UN Number 2875
Formula Weight 406.89
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hexachlorophene
EINECSNumber (4)-0088
RTECSNumber SM0700000
TSCA Yes