missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexachloroethane 99.0+%, TCI America™
$34.66 - $155.93
Chemical Identifiers
| CAS | 67-72-1 |
|---|---|
| Molecular Formula | C2Cl6 |
| Molecular Weight (g/mol) | 236.72 |
| MDL Number | MFCD00000799 |
| InChI Key | VHHHONWQHHHLTI-UHFFFAOYSA-N |
| Synonym | hexachloroethane, perchloroethane, ethane, hexachloro, avlothane, carbon hexachloride, hexachlorethane, mottenhexe, distokal, distopan, distopin |
| PubChem CID | 6214 |
| ChEBI | CHEBI:39227 |
| IUPAC Name | hexachloroethane |
| SMILES | ClC(Cl)(Cl)C(Cl)(Cl)Cl |
Chemical Identifiers
| 67-72-1 | |
| 236.72 | |
| VHHHONWQHHHLTI-UHFFFAOYSA-N | |
| 6214 | |
| hexachloroethane |
| C2Cl6 | |
| MFCD00000799 | |
| hexachloroethane, perchloroethane, ethane, hexachloro, avlothane, carbon hexachloride, hexachlorethane, mottenhexe, distokal, distopan, distopin | |
| CHEBI:39227 | |
| ClC(Cl)(Cl)C(Cl)(Cl)Cl |
Specifications
| 67-72-1 | |
| White | |
| MFCD00000799 | |
| 3077 | |
| VHHHONWQHHHLTI-UHFFFAOYSA-N | |
| hexachloroethane | |
| 6214 | |
| 236.72 | |
| Crystalline Powder |
| 185°C | |
| C2Cl6 | |
| 25 g | |
| hexachloroethane, perchloroethane, ethane, hexachloro, avlothane, carbon hexachloride, hexachlorethane, mottenhexe, distokal, distopan, distopin | |
| ClC(Cl)(Cl)C(Cl)(Cl)Cl | |
| 236.72 | |
| CHEBI:39227 | |
| ≥99.0% (GC) | |
| Hexachloroethane |
Safety and Handling
EINECSNumber : (2)-0057
RTECSNumber : KI4025000
TSCA : Yes