Learn More
Hexachloroethane, 99%
CAS: 67-72-1 | C2Cl6 | 236.72 g/mol
$54.47 - $54.47
Chemical Identifiers
| CAS | 67-72-1 |
|---|---|
| Molecular Formula | C2Cl6 |
| Molecular Weight (g/mol) | 236.72 |
| MDL Number | MFCD00000799 |
| InChI Key | VHHHONWQHHHLTI-UHFFFAOYSA-N |
| Synonym | hexachloroethane, perchloroethane, ethane, hexachloro, avlothane, carbon hexachloride, hexachlorethane, mottenhexe, distokal, distopan, distopin |
| PubChem CID | 6214 |
| ChEBI | CHEBI:39227 |
| SMILES | ClC(Cl)(Cl)C(Cl)(Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC148541000
|
Thermo Scientific Chemicals
148541000 |
100 g | Plastic bottle |
Each for $54.47
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
| 67-72-1 | |
| 236.72 | |
| VHHHONWQHHHLTI-UHFFFAOYSA-N | |
| 6214 | |
| ClC(Cl)(Cl)C(Cl)(Cl)Cl |
| C2Cl6 | |
| MFCD00000799 | |
| hexachloroethane, perchloroethane, ethane, hexachloro, avlothane, carbon hexachloride, hexachlorethane, mottenhexe, distokal, distopan, distopin | |
| CHEBI:39227 |
Specifications
| 67-72-1 | |
| White | |
| Authentic | |
| Plastic bottle | |
| Cl3CCCl3 | |
| 100 g | |
| 2.091 | |
| hexachloroethane, perchloroethane, ethane, hexachloro, avlothane, carbon hexachloride, hexachlorethane, mottenhexe, distokal, distopan, distopin | |
| VHHHONWQHHHLTI-UHFFFAOYSA-N | |
| hexachloroethane | |
| 6214 | |
| 236.74 | |
| Crystalline Powder or Crystals |
| 184.0°C to 190.0°C | |
| 2.0910g/mL | |
| 98.5% min. (GC) | |
| C2Cl6 | |
| MFCD00000799 | |
| 01,87 | |
| 15,4715 | |
| Solubility in water: 0.05g/L (22°C). Other solubilities: soluble in most common organic solvents,>50g/100mL ethanol | |
| ClC(Cl)(Cl)C(Cl)(Cl)Cl | |
| 236.72 | |
| CHEBI:39227 | |
| 99% | |
| Hexachloroethane |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN
GHS Signal Word: Warning
EINECSNumber : 200-666-4
RUO – Research Use Only