missing translation for 'onlineSavingsMsg'
Learn More

Hexachloroacetone 98.0+%, TCI America™

Catalog No. p-7130492
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H033525G 25 g
H0335500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H033525G Supplier TCI America Supplier No. H033525G
Only null left

Chemical Identifiers

CAS 116-16-5
Molecular Formula C3Cl6O
Molecular Weight (g/mol) 264.73
MDL Number MFCD00000796
InChI Key DOJXGHGHTWFZHK-UHFFFAOYSA-N
Synonym hexachloroacetone, hexachloro-2-propanone, perchloroacetone, hexachloropropanone, kureha hca, hca weedkiller, perchloro-2-propanone, bis trichloromethyl ketone, acetone, hexachloro, 2-propanone, hexachloro
PubChem CID 8303
ChEBI CHEBI:82243
IUPAC Name hexachloropropan-2-one
SMILES ClC(Cl)(Cl)C(=O)C(Cl)(Cl)Cl

Specifications

Color Colorless
Boiling Point 202°C
Quantity 25 g
UN Number 2661
Formula Weight 264.73
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexachloroacetone
EINECSNumber (2)-2341
RTECSNumber UC2100000
TSCA Yes