missing translation for 'onlineSavingsMsg'
Learn More

Hexabromostearic Acid 93.0+%, TCI America™

Catalog No. H005225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H005225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H005225G Supplier TCI America Supplier No. H005225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4167-08-2
Molecular Formula C18H30Br6O2
Molecular Weight (g/mol) 757.86
MDL Number MFCD00021795
InChI Key XIVWIZLTAYDHDS-UHFFFAOYNA-N
Synonym Hexabromooctadecanoic Acid
PubChem CID 95653
IUPAC Name 9,10,12,13,15,16-hexabromooctadecanoic acid
SMILES CCC(C(CC(C(CC(C(CCCCCCCC(=O)O)Br)Br)Br)Br)Br)Br

Specifications

Melting Point 178°C
Quantity 25 g
Formula Weight 757.86
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hexabromostearic Acid