accessibility menu, dialog, popup

UserName

CAS RN 4167-08-2

IUPAC Name: 9,10,12,13,15,16-hexabromooctadecanoic acid
Synonyms: 9,10,12,13,15,16-hexabromooctadecanoic acid
Molecular Weight (g/mol): 757.86
Molecular Formula: C18H30Br6O2
InChi Key: XIVWIZLTAYDHDS-UHFFFAOYNA-N
SMILES: CCC(Br)C(Br)CC(Br)C(Br)CC(Br)C(Br)CCCCCCCC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results