missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Hexabromocyclododecane, ≥97% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 134237-51-7 |
| Linear Formula | C12H18 Br6 |
| Melting Point | 152°C to 158°C |
| Synonym | (1 RS,2 SR,5 RS,6 RS,9 RS,10 SR)-1,2,5,6,9,10-Hexabromocyclododecane; (+/-)-beta-HBCD |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 641.70 |
| Percent Purity | ≥97% (HPLC) |
| Grade | Analytical Standard |