missing translation for 'onlineSavingsMsg'
Learn More

Hexaamminecobalt(III) Chloride 99.0+%, TCI America™

Catalog No. p-7130313
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H005125G 25 g
H0051500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H005125G Supplier TCI America Supplier No. H005125G
Only null left

Chemical Identifiers

CAS 10534-89-1
Molecular Formula H18Cl3CoN6
MDL Number MFCD00036304
Synonym Luteo-cobaltic Chloride

Specifications

Physical Form Crystalline Powder
Quantity 25 g
UN Number 2811
Formula Weight 267.47
Percent Purity ≥99.0% (T)
Chemical Name or Material Hexaamminecobalt(III) Chloride