missing translation for 'onlineSavingsMsg'
Learn More

Hexaamminecobalt(III) chloride, 99%

Catalog No. p-7022610
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1547014 25 g
AAA1547022 100 g
AAA1547036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1547014 Supplier Thermo Scientific Chemicals Supplier No. A1547014
Only null left

CAS: 10534-89-1 | H18Cl3CoN6

Hexaamminecobalt(III) chloride is a component of structural biology methods, especially for DNA or RNA, where the positive ions stabilize the tertiary structure of the phosphate backbone, which favors to solve their structure by X-ray crystallography. It serves as an excellent antibacterial and antiviral property.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaamminecobalt(III) chloride is a component of structural biology methods, especially for DNA or RNA, where the positive ions stabilize the tertiary structure of the phosphate backbone, which favors to solve their structure by X-ray crystallography. It serves as an excellent antibacterial and antiviral property.

Solubility
Soluble in ammonia and water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 10534-89-1
Molecular Formula H18Cl3CoN6
MDL Number MFCD00036304

Specifications

Density 1.71
Odor Odorless
Linear Formula [Co(NH3)6]Cl3
Quantity 25 g
Merck Index 14,4675
Solubility Information Soluble in ammonia and water.
Formula Weight 267.48
Percent Purity 99%
Chemical Name or Material Hexaamminecobalt(III) chloride
Hazard Category H317-H334-H350
Hazard Statement GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
EINECSNumber 234-103-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only