missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Hex Recombinant Rabbit Monoclonal Antibody (23GB6630)
GREENER_CHOICE

Catalog No. MA553082
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553082 100 μL
MA553083 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553082 Supplier Invitrogen™ Supplier No. MA553082
Only null left

Rabbit Recombinant Monoclonal Antibody

Hex Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

HHEX is a member of the homeobox family of transcription factors, many of which are involved in developmental processes. Expression in specific hematopoietic lineages suggests that this protein may play a role in hematopoietic differentiation. Establishes anterior identity at two levels 1) acts early to enhance canonical WNT-signaling by repressing expression of TLE4 and 2) acts later to inhibit NODAL-signaling by directly targeting NODAL.

Specifications

Antigen Hex
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6630
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HHEX
Gene Accession No. Q03014
Gene Alias hematopoietically expressed homeobox; hematopoietically expressed homeobox, related sequence 2; hematopoietically-expressed homeobox protein Hhex; HEX; Hex1; HHEX; Hhex-rs2; HMPH; homeobox; homeobox protein HEX; Homeobox protein PRH; homeobox, hematopoietically expressed; HOX11L-PEN; mHex; Prh; Prhx; proline-rich homeodomain-containing transcription factor
Gene Symbols HHEX
Host Species Rabbit
Immunogen A synthesized peptide derived from human Hex (100-133AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3087
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less