Learn More
Applied Biosystems™ HEX™ Dye Phosphoramidite
Contains 105 mg HEX™ Dye Phosphoramidite .HEX™ Dye Phosphoramidite is used for the 5’ attachment of HEX™ dye during oligonucleotide synthesis.The label is treated as if it were simply another base: It is inserted at the appropriate place in the sequence and incorporated automatically.
Supplier:  Applied Biosystems™ 401526
Encompass
 View more versions of this product
Catalog No. 401526
Only null left
Description
Description
On-instrument labeling typically saves hours of post-synthesis manual processingFluorescent label HEX™ is a single-isomer fluorescein dye designed for detection and analysis of labeled PCR products on ABI PRISM™ genetic analysis systems. HEX™ is equally useful in applications historically employing isotopic labels. As such, it eliminates the hazard associated with handling, storage, and disposal of radioactive materials.
  • Specialty phosphoramidites allow online addition of the label during synthesis, saving time in production of labeled probes and primers.
  • Designed and optimized for use with ABI synthesizers, guaranteeing reliable nucleic acid synthesis.
  • We offer a range of specialty phosphoramites and linkers to give you the flexibility demanded in oligonucleotide synthesis.
Specifications
Specifications
HEX Dye Phosphoramidite
Shipped on Dry Ice
105 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.