missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ HEX™ Dye Phosphoramidite

Catalog No. p-7083026
Encompass
Change view
Click to view available options
Quantity:
55 mg
105 mg
Product Type:
HEX Dye CEP
HEX Dye Phosphoramidite
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
403170 55 mg HEX Dye CEP
401526 105 mg HEX Dye Phosphoramidite
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 403170 Supplier Applied Biosystems™ Supplier No. 403170
Only null left

Contains 105 mg HEX™ Dye Phosphoramidite .HEX™ Dye Phosphoramidite is used for the 5’ attachment of HEX™ dye during oligonucleotide synthesis.The label is treated as if it were simply another base: It is inserted at the appropriate place in the sequence and incorporated automatically.

On-instrument labeling typically saves hours of post-synthesis manual processingFluorescent label HEX™ is a single-isomer fluorescein dye designed for detection and analysis of labeled PCR products on ABI PRISM™ genetic analysis systems. HEX™ is equally useful in applications historically employing isotopic labels. As such, it eliminates the hazard associated with handling, storage, and disposal of radioactive materials.
  • Specialty phosphoramidites allow online addition of the label during synthesis, saving time in production of labeled probes and primers
  • Designed and optimized for use with ABI™ synthesizers, guaranteeing reliable nucleic acid synthesis
  • We offer a range of specialty phosphoramites and linkers to give you the flexibility demanded in oligonucleotide synthesis

Specifications

Detection Method Fluorescence
Format Powder
For Use With (Equipment) 3900 DNA Synthesizer
Product Type HEX Dye CEP
Content And Storage This reagent should be stored dessicated at -20°C.
Nucleoside Type None (Modifier)
Shipping Condition Dry Ice
Sample Type DNA
Quantity 55 mg
Reagent Type Phosphoramidite
Nucleic Acid Type None (Modifier)
Product Line HEX
For Use With (Application) Oligo Synthesis
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.