missing translation for 'onlineSavingsMsg'
Learn More

Hesperidin 90.0+%, TCI America™

Catalog No. p-7130311
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H004925G 25 g
H0049100G 100 g
H0049500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H004925G Supplier TCI America Supplier No. H004925G
Only null left

Chemical Identifiers

CAS 520-26-3
Molecular Formula C28H34O15
Molecular Weight (g/mol) 610.57
MDL Number MFCD00075663
InChI Key QUQPHWDTPGMPEX-QJBIFVCTSA-N
Synonym Vitamin P
PubChem CID 45358131
IUPAC Name (2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-3,4-dihydro-2H-1-benzopyran-4-one
SMILES COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1

Specifications

Melting Point 254°C
Color Yellow
Quantity 25 g
Formula Weight 610.57
Percent Purity ≥90.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Hesperidin
RTECSNumber MK6650000
TSCA Yes