missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Hesperidin, 90+%
CAS: 520-26-3 | C28H34O15 | 610.57 g/mol
$35.02 - $35.02
Chemical Identifiers
| CAS | 520-26-3 |
|---|---|
| Molecular Formula | C28H34O15 |
| Molecular Weight (g/mol) | 610.57 |
| MDL Number | MFCD00075663 |
| InChI Key | QUQPHWDTPGMPEX-QJBIFVCTSA-N |
| PubChem CID | 45358131 |
| SMILES | COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC123460050
|
Thermo Scientific Chemicals
123460050 |
5 g | Glass Bottle |
Each for $35.02
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 520-26-3 | |
| 610.57 | |
| QUQPHWDTPGMPEX-QJBIFVCTSA-N | |
| COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1 |
| C28H34O15 | |
| MFCD00075663 | |
| 45358131 |
Specifications
| 520-26-3 | |
| Brown to White | |
| Authentic | |
| Glass Bottle | |
| MFCD00075663 | |
| 15, 4706 | |
| Solubility in water: 1g/50l. Other solubilities: soluble in diluted alkalies,soluble in pyridine,soluble in formamide and dimethylformamide (60 c),slightly sol. in methanol and glacial acetic acid,practically insol. in acetone benzene and chcl3 | |
| COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1 | |
| 610.57 | |
| 610.54 | |
| Powder |
| 258.0°C to 262.0°C | |
| 4% max. | |
| 97% | |
| C28H34O15 | |
| 5 g | |
| − 76.00 | |
| QUQPHWDTPGMPEX-QJBIFVCTSA-N | |
| (2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-3,4-dihydro-2H-1-benzopyran-4-one | |
| 45358131 | |
| 97% | |
| Hesperidin, 97% |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 208-288-1
RTECSNumber : MK6650000
TSCA : TSCA
RUO – Research Use Only