missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Hesperidin, 90+%
Supplier: Thermo Scientific™ 123462500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Hesperidin | |
| 520-26-3 | |
| Authentic | |
| Plastic bottle | |
| 250g | |
| 15, 4706 | |
| Solubility in water: 1g/50l. Other solubilities: soluble in diluted alkalies,soluble in pyridine,soluble in formamide and dimethylformamide (60 c),slightly sol. in methanol and glacial acetic acid,practically insol. in acetone benzene and chcl3 | |
| COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1 | |
| 610.57 | |
| 610.54 |
| 97% | |
| 4% max. | |
| 97% | |
| C28H34O15 | |
| MFCD00075663 | |
| − 76.00 | |
| QUQPHWDTPGMPEX-QJBIFVCTSA-N | |
| (2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxy-2,3-dihydrochromen-4-one | |
| 45358131 | |
| 97% |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 208-288-1
RTECSNumber : MK6650000
TSCA : TSCA