missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hesperetin, MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| CAS | 69097-99-0 |
| Physical Form | Neat |
| Linear Formula | C16H14O6 |
| MDL Number | MFCD00016956 |
| Synonym | 3′,5,7-Trihydroxy-4′-methoxyflavanone; 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4-chromanone |
| Type | Analytical Standard |
| Quantity | 10 g |
| Formula Weight | 302.28 |
| Grade | Analytical Standard |